2,6-Dimethylphenol, 99%
2,6-Dimethylphenol, 99%
2,6-Dimethylphenol, 99%
Thermo Scientific Chemicals

2,6-Dimethylphenol, 99%

CAS: 576-26-1 | C8H10O | 122.17 g/mol
Quantity:
500 g
2500 g
Catalog number A11947.36
also known as A11947-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS576-26-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to yellow-brown or orange or colorless
FormCrystals or crystalline powder or fused solid or low melting solid
Assay (GC)≥98.5% (non-U.S. specification)
Assay≥98.5% (U.S. specification)
Water Content (Karl Fischer Titration)<2.0% (non-U.S. specification)
View more
2,6-dimethylphenol is used in the synthesis of anti-oxidant compounds due to the phenol moiety in the structure. In addition, this compound is used as a reactant in the synthesis of polyphenylene ether polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-dimethylphenol is used in the synthesis of anti-oxidant compounds due to the phenol moiety in the structure. In addition, this compound is used as a reactant in the synthesis of polyphenylene ether polymers.

Solubility
Soluble in water and chloroform.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
RUO – Research Use Only

General References:

  1. Yasar Thewalim; Fredrik Aldaeus; Anders Colmsjö. Retention time prediction of compounds in Grob standard mixture for apolar capillary columns in temperature-programmed gas chromatography. Analytical and Bioanalytical Chemistry.2009,393(1), 327-334.
  2. Masahiro Ogata et. al. Antioxidant activity of propofol and related monomeric and dimeric compounds. Chemical & Pharmaceutical Bulletin,2005, 53(3), 344-346.
  3. O-Alkylation of this hindered phenol by alkyl halides has been achieved using NaOH with micellar catalysis by (1-Hexadecyl) trimethyl ammonium bromide, A15235: Tetrahedron, 44, 6677 (1988). Alkylation of the Li salt with MeI occurs preferentially on carbon to give the dimer of 2,6,6-trimethyl-2,4-cyclohexadienone: Org. Synth. Coll., 5, 1092 (1973).
  4. Addition of lithiated 2,6-dimethylphenyl esters of alkanoic acids to carbonyl compounds has been used in a highly diastereoselective synthesis of ß-hydroxy acids. In some cases, only one of the two possible diastereomers is formed. For list of examples, see: Org. Synth. Coll., 7, 190 (1990).