1-Bromo-2,4-dichlorobenzene, 98%
1-Bromo-2,4-dichlorobenzene, 98%
1-Bromo-2,4-dichlorobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-2,4-dichlorobenzene, 98%

CAS: 1193-72-2 | C6H3BrCl2 | 225.894 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A12298.14
also known as A12298-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1193-72-2
IUPAC Name1-bromo-2,4-dichlorobenzene
Molecular FormulaC6H3BrCl2
InChI KeyISHYFWKKWKXXPL-UHFFFAOYSA-N
SMILESClC1=CC=C(Br)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5965-1.6005 @ 20?C
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4'-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4′-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jeanne L. Bollige.; Christian M. Frech. The 1,3-Diaminobenzene-Derived Aminophosphine Palladium Pincer Complex {C6H3[NHP(piperidinyl)2]2Pd(Cl)} - A Highly Active Suzuki-Miyaura Catalyst with Excellent Functional Group Tolerance. Advanced Synthesis & Catalysis. 2010, 352 (6), 1075-1080.
  2. Okada, Y.; Yokozawa, M.;Akiba, M., Oishi, K.; Kyoji, O.; Akeboshi, T.; Nishimura, J. Bromination by means of sodium monobromoisocyanurate (SMBI). Organic & biomolecular chemistry. 2003, 1 (14), 2506-2511.