3-Bromo-4-methoxybenzoic acid, 98+%
3-Bromo-4-methoxybenzoic acid, 98+%
3-Bromo-4-methoxybenzoic acid, 98+%
Thermo Scientific Chemicals

3-Bromo-4-methoxybenzoic acid, 98+%

CAS: 99-58-1 | C8H6BrO3 | 230.04 g/mol
Quantity:
5 g
25 g
Catalog number A12653.14
also known as A12653-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS99-58-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)216.5-222.5?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only