2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%
2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%
2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%

CAS: 60706-63-0 | C10H13ClO2S | 232.72 g/mol
Quantity:
10 g
50 g
Catalog number A13313.18
also known as A13313-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS60706-63-0
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)95.0-102.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only