2-Sulfobenzoic anhydride, 94%
2-Sulfobenzoic anhydride, 94%
2-Sulfobenzoic anhydride, 94%
Thermo Scientific Chemicals

2-Sulfobenzoic anhydride, 94%

CAS: 81-08-3 | C7H4O4S | 184.165 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A13314.14
also known as A13314-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS81-08-3
IUPAC Name3H-2,1λ⁶-benzoxathiole-1,1,3-trione
Molecular FormulaC7H4O4S
InChI KeyNCYNKWQXFADUOZ-UHFFFAOYSA-N
SMILESO=C1OS(=O)(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or granules
Assay (Titration ex Anhydride)≥92.0 to ≤108.0%
Melting Point (clear melt)119.5-128.5?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only