3',4'-Dihydroxypropiophenone, 98%
3',4'-Dihydroxypropiophenone, 98%
3',4'-Dihydroxypropiophenone, 98%
Thermo Scientific Chemicals

3',4'-Dihydroxypropiophenone, 98%

CAS: 7451-98-1 | C9H10O3 | 166.176 g/mol
Quantity:
1 g
5 g
Catalog number A13444.06
also known as A13444-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7451-98-1
IUPAC Name1-(3,4-dihydroxyphenyl)propan-1-one
Molecular FormulaC9H10O3
InChI KeyHNWIHBDMOYWCGX-UHFFFAOYSA-N
SMILESCCC(=O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)145.0-151.0?C
Assay (GC)≥97.5%
Appearance (Color)Grey or pale grey/brown
FormPowder
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Antonio Trincone; Edoardo Pagnotta. Efficient chemoselective synthesis of 3,4'-dihydroxypropiophenone 3-O-β-D-glucoside by thermophilic β-glycosidase from Sulfolobus solfataricus. Biotechnology Letters. 1995, 17 (1), 45-48.
  2. Elwin E. Harris; James D'Ianni; Homer Adkins. Reaction of Hardwood Lignin with Hydrogen. J. Am. Chem. Soc, 1938, 60 (6), 1467-1470.