Naphthalene-2-sulfonic acid hydrate, 94%
Naphthalene-2-sulfonic acid hydrate, 94%
Naphthalene-2-sulfonic acid hydrate, 94%
Thermo Scientific Chemicals

Naphthalene-2-sulfonic acid hydrate, 94%

CAS: 76530-12-6 | C10H8O3S | 208.23 g/mol
Quantity:
250 g
1000 g
Catalog number A13445.0B
also known as A13445-0B
Price (JPY)
-
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS76530-12-6
IUPAC Namenaphthalene-2-sulfonic acid
Molecular FormulaC10H8O3S
InChI KeyKVBGVZZKJNLNJU-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink to pale gray to gray
Assay (Aqueous acid-base Titration)≥92.0 to ≤108.0%
Water Content (Karl Fischer Titration)≤17.78%
FormCrystals or powder or crystalline powder
Assay (HPLC)≥92.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only