3-Amino-1H-pyrazole-4-carbonitrile, 98%
3-Amino-1H-pyrazole-4-carbonitrile, 98%
3-Amino-1H-pyrazole-4-carbonitrile, 98%
Thermo Scientific Chemicals

3-Amino-1H-pyrazole-4-carbonitrile, 98%

CAS: 16617-46-2 | C4H4N4 | 108.104 g/mol
Quantity:
5 g
25 g
Catalog number A14266.06
also known as A14266-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS16617-46-2
IUPAC Name5-amino-1H-pyrazole-4-carbonitrile
Molecular FormulaC4H4N4
InChI KeyFFNKBQRKZRMYCL-UHFFFAOYSA-N
SMILESNC1=C(C=NN1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to orange-brown
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)171-178°C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only