3,5-Dimethylbenzyl bromide, 98%
3,5-Dimethylbenzyl bromide, 98%
3,5-Dimethylbenzyl bromide, 98%
Thermo Scientific Chemicals

3,5-Dimethylbenzyl bromide, 98%

CAS: 27129-86-8 | C9H11Br | 199.091 g/mol
Quantity:
5 g
25 g
Catalog number A14267.14
also known as A14267-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS27129-86-8
IUPAC Name1-(bromomethyl)-3,5-dimethylbenzene
Molecular FormulaC9H11Br
InChI KeyQXDHXCVJGBTQMK-UHFFFAOYSA-N
SMILESCC1=CC(CBr)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale cream to pale yellow
FormCrystals or powder or crystalline powder or clear liquid as melt
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)34.0-41.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only