Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals

Quantity:
10 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14268.22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS756-79-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥ 96.0%
FormLiquid
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

Solubility
Miscible with water, ethanol, ether, alcohol, benzene, acetone and carbon tetrachloride. Immiscible with heavy mineral oil.

Notes
Incompatible materials strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. The lithio-derivative reacts with esters to give ß-ketophosphonates, useful precursors of ɑß-unsaturated ketones by Horner-Wadsworth-Emmons olefination: J. Am. Chem. Soc., 88, 5654 (1966). An intramolecular version leads to cyclopentenones: Synth. Commun., 5, 1 (1975); Tetrahedron Lett., 22, 257 (1981). Dimethyl glutarate gives the 3-(phosphonatomethyl)cyclohexenone, olefination of which affords the corresponding 3-alkenylcycloalkenone. Dimethyl succinate also gives this reaction, but in lower yield; higher diesters give acyclic products: J. Org. Chem., 59, 1943 (1994):
  2. See also Diethyl methyl phosphonate, A14772 and Appendix 1.
  3. Park, E. J.; Han, S. W.; Jeong, B.; Park, S. H.; Kim, Y. G.; Kim, Y. H.; Kim, Y. D. Effect of polydimethylsiloxane (PDMS) coating on TiO2-based MALDI matrix for dimethyl methylphosphonate (DMMP) analysis. Appl. Surf. Sci. 2015, 353, 342-349.
  4. Yoo, R.; Cho, S.; Song, M. J.; Lee, W. Highly sensitive gas sensor based on Al-doped ZnO nanoparticles for detection of dimethyl methylphosphonate as a chemical warfare agent simulant. Sens. Actuators, B 2015, 221, 217-223.