Benzyltriethylammonium bromide, 98+%
Benzyltriethylammonium bromide, 98+%
Benzyltriethylammonium bromide, 98+%
Thermo Scientific Chemicals

Benzyltriethylammonium bromide, 98+%

CAS: 5197-95-5 | C13H22BrN | 272.23 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A14291.14
also known as A14291-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5197-95-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
Appearance (Color)White to pale cream to pale yellow
Water Content (Karl Fischer Titration)≤1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only