4-tert-Butylbenzoyl chloride, 98%
4-tert-Butylbenzoyl chloride, 98%
4-tert-Butylbenzoyl chloride, 98%
Thermo Scientific Chemicals

4-tert-Butylbenzoyl chloride, 98%

CAS: 1710-98-1 | C11H13ClO | 196.67 g/mol
Quantity:
25 g
100 g
Catalog number A14373.14
also known as A14373-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1710-98-1
IUPAC Name4-tert-butylbenzoyl chloride
Molecular FormulaC11H13ClO
InChI KeyWNLMYNASWOULQY-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5340-1.5380 @ 20?C
4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9?-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9′-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. Yuh-Yih Chien et. al. Syntheses and spectroscopic studies of spirobifluorene-bridged bipolar systems; photoinduced electron transfer reactions.Chem Commun (Camb).,2002,7(23), 2874-5.
  2. Jiang X, et al. Statistical copolymers with side-chain hole and electron transport groups for single-layer electroluminescent device applications.Chem. Mater.,2000,12(9), 2542-2549.