Methyl 3-hydroxybenzoate, 99%
Methyl 3-hydroxybenzoate, 99%
Methyl 3-hydroxybenzoate, 99%
Thermo Scientific Chemicals

Methyl 3-hydroxybenzoate, 99%

CAS: 19438-10-9 | C8H8O3 | 152.15 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A14425.14
also known as A14425-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS19438-10-9
IUPAC Namemethyl 3-hydroxybenzoate
Molecular FormulaC8H8O3
InChI KeyYKUCHDXIBAQWSF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale gray
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)67.0-75.0°C
Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

Solubility
Soluble in chloroform, dimetyl Sulfoxide,ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Alvarez-Rivera, G.; Llompart, M.; Garcia-Jares, C.; Lores, M. Identification of unwanted photoproducts of cosmetic preservatives in personal care products under ultraviolet-light using solid-phase microextraction and micro-matrix solid-phase dispersion. J. Chromatogr. A 2015, 1390, 1-12.
  2. Banerjee, U.; DeBerardinis, A. M.; Hadden, M. K. Design, synthesis, and evaluation of hybrid vitamin D3 side chain analogues as hedgehog pathway inhibitors. Bioorg. Med. Chem. 2015, 23 (3), 548-555.