2-Heptyn-1-ol, 97%
2-Heptyn-1-ol, 97%
2-Heptyn-1-ol, 97%
Thermo Scientific Chemicals

2-Heptyn-1-ol, 97%

CAS: 1002-36-4 | C7H12O | 112.172 g/mol
Quantity:
5 g
25 g
Catalog number A14959.14
also known as A14959-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1002-36-4
IUPAC Namehept-2-yn-1-ol
Molecular FormulaC7H12O
InChI KeyOGDYROFIIXDTQK-UHFFFAOYSA-N
SMILESCCCCC#CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4525-1.4575 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only