(+/-)-Camphorquinone, 99%
(+/-)-Camphorquinone, 99%
(+/-)-Camphorquinone, 99%
(+/-)-Camphorquinone, 99%
Thermo Scientific Chemicals

(+/-)-Camphorquinone, 99%

CAS: 10373-78-1 | C10H14O2 | 166.22 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A14967.14
also known as A14967-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10373-78-1
IUPAC Name1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Molecular FormulaC10H14O2
InChI KeyVNQXSTWCDUXYEZ-UHFFFAOYNA-N
SMILESCC1(C)C2CCC1(C)C(=O)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)199-206?C
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

Solubility
Soluble in ethanol, ethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ratanajanchai, M.; Tanwilai, D.; Sunintaboon, P. Visible light-induced surfactant-free emulsion polymerization using camphorquinone/tertiary amine as the initiating system for the synthesis of amine-functionalized colloidal nanoparticles. J. Colloid Interface Sci. 2013, 409, 25-31.
  2. Wessels, M.; Leyhausen, G.; Volk, J.; Geurtsen, W. Oxidative stress is responsible for genotoxicity of camphorquinone in primary human gingival fibroblasts. Clin.Oral.Invest. 2014, 18 (6), 1705-1710.