Barium thiosulfate, 98%
Barium thiosulfate, 98%
Barium thiosulfate, 98%
Thermo Scientific Chemicals

Barium thiosulfate, 98%

CAS: 35112-53-9 | BaO3S2 | 249.444 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A14981.14
also known as A14981-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS35112-53-9
IUPAC Namebarium(2+) 1-oxo-1λ⁶-disulfene-1,1-bis(olate)
Molecular FormulaBaO3S2
InChI KeyONPIOWQPHWNPOQ-UHFFFAOYSA-L
SMILES[Ba++].[O-]S([O-])(=O)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofA≥97.5% to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only