Ethyl 3-hydroxybenzoate, 99%
Ethyl 3-hydroxybenzoate, 99%
Ethyl 3-hydroxybenzoate, 99%
Ethyl 3-hydroxybenzoate, 99%
Thermo Scientific Chemicals

Ethyl 3-hydroxybenzoate, 99%

CAS: 7781-98-8 | C9H10O3 | 166.176 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A15343.36
also known as A15343-36
Price (JPY)
-
Request A Quote
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7781-98-8
IUPAC Nameethyl 3-hydroxybenzoate
Molecular FormulaC9H10O3
InChI KeyMWSMNBYIEBRXAL-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)69.0-76.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction. It is a food-grade antimicrobial agent.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Marta Goretti; Benedetta Turchetti; Morena Buratta; Eva Branda; Lanfranco Corazzi; Ann Vaughan-Martini; Pietro Buzzini. In vitro antimycotic activity of aWilliopsis saturnuskiller protein against food spoilage yeasts. International Journal of Food Microbiology. 2009, 131,(2), 178-182.
  2. S.Ren. Determining the mechanisms of toxic action of phenols to Tetrahymena pyriformis. Environmental Toxicology. 2002, 17,(2), 119-127.