2-Chloro-4-fluorobenzaldehyde, 97%
2-Chloro-4-fluorobenzaldehyde, 97%
2-Chloro-4-fluorobenzaldehyde, 97%
Thermo Scientific Chemicals

2-Chloro-4-fluorobenzaldehyde, 97%

CAS: 84194-36-5 | C7H4ClFO | 158.56 g/mol
Quantity:
1 g
5 g
25 g
Catalog number A15345.14
also known as A15345-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS84194-36-5
IUPAC Name2-chloro-4-fluorobenzaldehyde
Molecular FormulaC7H4ClFO
InChI KeyKMQWNQKESAHDKD-UHFFFAOYSA-N
SMILESFC1=CC=C(C=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or chunks
Free acid (titration)≤1.5%
Melting Point (clear melt)55.0-65.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only