Lithium hydroxide monohydrate, 98%
Lithium hydroxide monohydrate, 98%
Lithium hydroxide monohydrate, 98%
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, 98%

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol
Quantity:
100 g
500 g
2500 g
Catalog number A15519.36
also known as A15519-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

Solubility
Soluble in water, methanol and ethanol.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture, strong oxidizing agents, strong acids and carbon dioxide.
RUO – Research Use Only

General References:

  1. Preferred base for chemoselective protection of carboxylic acids as methyl esters by reaction with dimethyl sulfate in THF, as an alternative to diazomethane: J. Org. Chem., 64, 8014 (1999).
  2. Noda, Y.; Koga, N. Phenomenological kinetics of the carbonation reaction of lithium hydroxide monohydrate: Role of surface product layer and possible existence of a liquid phase. J. Phys. Chem. C 2014, 118 (10), 5424-5436.
  3. Wang, D.; Belharouak, I.; Ortega, L. H.; Zhang, X.; Rui Xu, R.; Zhou, D.; Zhou, G.; Amine, K. Synthesis of high capacity cathodes for lithium-ion batteries by morphology-tailored hydroxide co-precipitation. J. Power Sources 2015, 274, 451-457.