4-Fluoroaniline, 99%
4-Fluoroaniline, 99%
4-Fluoroaniline, 99%
Thermo Scientific Chemicals

4-Fluoroaniline, 99%

CAS: 371-40-4 | C6H6FN | 111.12 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A15597.36
also known as A15597-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS371-40-4
IUPAC Name4-fluoroaniline
Molecular FormulaC6H6FN
InChI KeyKRZCOLNOCZKSDF-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear yellow to orange to brown
Refractive Index1.5380-1.5410 @ 20?C
Assay (GC)≥98.5%
It finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.

Solubility
Slightly soluble in water. Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Scarfe GB, et al. Studies on the metabolism of 4-fluoroaniline and 4-fluoroacetanilide in rat: formation of 4-acetamidophenol (paracetamol) and its metabolites via defluorination and N-acetylation.Xenobiotica.,1999,29(2), 205-216.
  2. Song E, et al. Isolation, identification and characterization of a novel Ralstonia sp. FD-1, capable of degrading 4-fluoroaniline.Biodegradation.,2014,25(1), 85-94.