Bis(trimethylsilyl)methane, 98%
Bis(trimethylsilyl)methane, 98%
Bis(trimethylsilyl)methane, 98%
Thermo Scientific Chemicals

Bis(trimethylsilyl)methane, 98%

CAS: 2117-28-4 | C7H20Si2 | 160.41 g/mol
Quantity:
5 g
25 g
Catalog number A15662.06
also known as A15662-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2117-28-4
IUPAC Nametrimethyl[(trimethylsilyl)methyl]silane
Molecular FormulaC7H20Si2
InChI KeyGYIODRUWWNNGPI-UHFFFAOYSA-N
SMILESC[Si](C)(C)C[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Refractive Index1.4150-1.4190 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only