2,6-Dibromo-4-isopropylaniline, 98%
2,6-Dibromo-4-isopropylaniline, 98%
2,6-Dibromo-4-isopropylaniline, 98%
2,6-Dibromo-4-isopropylaniline, 98%
Thermo Scientific Chemicals

2,6-Dibromo-4-isopropylaniline, 98%

CAS: 10546-65-3 | C9H11Br2N | 293.002 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A15932.14
also known as A15932-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10546-65-3
IUPAC Name2,6-dibromo-4-(propan-2-yl)aniline
Molecular FormulaC9H11Br2N
InChI KeyCJEBZUFROMNDEK-UHFFFAOYSA-N
SMILESCC(C)C1=CC(Br)=C(N)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream or pale pink or pale green
Assay (GC)≥97.5%
FormPowder
Melting Point (clear melt)43.5-49.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only