Potassium acetate, 99%
Potassium acetate, 99%
Potassium acetate, 99%
Thermo Scientific Chemicals

Potassium acetate, 99%

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol
Quantity:
10,000 g
2500 g
Catalog number A16321.0C
also known as A16321-0C
Price (JPY)
-
Quantity:
10,000 g
Request bulk or custom format
Chemical Identifiers
CAS127-08-2
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (dry wt. basis)
Identification (FTIR)Conforms
Loss on Drying≤2.25%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Potassium acetate is used as deicer to remove snow and ice from the surface and considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive, as a buffer in biological research, as an extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins, thereby removal of proteins from DNA. It plays an important role in preserving tissues, fixation and mummification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium acetate is used as deicer to remove snow and ice from the surface and considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive, as a buffer in biological research, as an extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins, thereby removal of proteins from DNA. It plays an important role in preserving tissues, fixation and mummification.

Solubility
Soluble in water, alcohol and ammonia. Insoluble in acetone and ether.

Notes
Hygroscopic. Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Has been found to give faster reaction than sodium acetate in the Perkin condensation of carboxylic acid anhydrides with aromatic aldehydes: Org. Synth. Coll., 3, 127, 426 (1955); review: Org. React., 1, 210 (1942).
  2. The nucleophilicity of acetate anion, e.g. in benzene or acetonitrile, is greatly increased in the presence of 18-crown-6: Tetrahedron Lett., 2417 (1974). In a comparative study of various anions in acetonitrile, 'naked' acetate, formed in this manner, was surprisingly found to be the most nucleophilic: Tetrahedron Lett., 4205 (1975).
  3. Kathiravan, S.; Nicholls, I. A. Palladium Catalyzed Vinyltrifluoromethylation of Aryl Halides through Decarboxylative Cross-Coupling with 2-(Trifluoromethyl) acrylic Acid. Org. Lett. 2015, 17 (8), 1874-1877.
  4. Rodríguez-Palmeiro, I.; Rodríguez-Escontrela, I.; Rodríguez, O.; Arce, A.; Soto, A. Characterization and interfacial properties of the surfactant ionic liquid 1-dodecyl-3-methyl imidazolium acetate for enhanced oil recovery. RSC Adv. 2015, 5 (47), 37392-37398.