Dotriacontane, 98%
Dotriacontane, 98%
Dotriacontane, 98%
Thermo Scientific Chemicals

Dotriacontane, 98%

CAS: 544-85-4 | C32H66 | 450.88 g/mol
Quantity:
5 g
25 g
Catalog number A17480.14
also known as A17480-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS544-85-4
IUPAC Namedotriacontane
Molecular FormulaC32H66
InChI KeyQHMGJGNTMQDRQA-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or flakes
Appearance (Color)White
Assay (GC)>97.5%
Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.

Solubility
Soluble in hot toluene, diethyl ether, benzene and ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. González-Arias, S.; Verónico-Sánchez, F. J.; Elizalde-Solis, O.; Zúñiga-Moreno, A.; Camacho-Camacho, L. E. An enhanced first freezing point method for solid-liquid-gas equilibrium measurements in binary systems. J. Supercrit. Fluids 2015, 104, 301-306.
  2. Lawal, O. A.; Ogunwande, I. A.; Ibirogba, A. E.; Layode, O. M.; Opoku, A. R. Chemical Constituents of Essential Oils from Catharanthus roseus (L.) G. Don Grown in Nigeria. J Essent Oil Bear Pl 2015, 18 (1), 57-63.