N,N,N',N'-Tetraethyl-1,3-propanediamine, 97%
N,N,N',N'-Tetraethyl-1,3-propanediamine, 97%
N,N,N',N'-Tetraethyl-1,3-propanediamine, 97%
Thermo Scientific Chemicals

N,N,N',N'-Tetraethyl-1,3-propanediamine, 97%

CAS: 60558-96-5 | C11H26N2 | 186.343 g/mol
Quantity:
5 g
25 g
Catalog number A17680.14
also known as A17680-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS60558-96-5
IUPAC Name[3-(diethylazaniumyl)propyl]diethylazanium
Molecular FormulaC11H28N2
InChI KeyRCZLVPFECJNLMZ-UHFFFAOYSA-P
SMILESCC[NH+](CC)CCC[NH+](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4355-1.4405 @ 20°C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only