2-Pentene, cis + trans, 98%
2-Pentene, cis + trans, 98%
2-Pentene, cis + trans, 98%
2-Pentene, cis + trans, 98%
Thermo Scientific Chemicals

2-Pentene, cis + trans, 98%

CAS: 109-68-2 | C5H10 | 70.14 g/mol
Quantity:
25 g
100 g
Catalog number A17736.14
also known as A17736-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS109-68-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5% (sum of isomers)
FormLiquid
Refractive Index1.3795-1.3845 @ 20?C
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only

General References:

  1. Myers SB and Register RA. Synthesis of narrow-distribution polycyclopentene using a ruthenium ring-opening metathesis initiator.Polymer,2008,49(4), 1877-882.
  2. Bing-Jun Zhao.; Henry J Shine.; John N Marx.; Cristina Hofmann.; Kenton H Whitmire. Addition of the phenoxathiin cation radical to alkenes and nonconjugated dienes. Formation of (E)- and (Z)-(10-phenoxathiiniumyl)alkenes and (E)- and (Z)-(10-phenoxathiiniumyl)dienes on basic alumina.J Org Chem.2007,72(16), 6154-6161.