Thermo Scientific Chemicals

4,5-Imidazoledicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A17783.22
100 g, Each
Thermo Scientific Chemicals

4,5-Imidazoledicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A17783.22
100 g, Each
Quantity
Catalog number: A17783.22
also known as A17783-22
Price (JPY)

Chemical Identifiers

CAS
570-22-9
IUPAC Name
1H-imidazole-4,5-dicarboxylic acid
Molecular Formula
C5H4N2O4
InChI Key
ZEVWQFWTGHFIDH-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(N=CN1)C(O)=O
Appearance (Color)
White to cream
Acid-base back titration
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text