4-Chloro-3-methylphenol, 99%
4-Chloro-3-methylphenol, 99%
4-Chloro-3-methylphenol, 99%
Thermo Scientific Chemicals

4-Chloro-3-methylphenol, 99%

CAS: 59-50-7 | C7H7ClO | 142.582 g/mol
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17823.22
also known as A17823-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS59-50-7
IUPAC Name4-chloro-3-methylphenol
Molecular FormulaC7H7ClO
InChI KeyCFKMVGJGLGKFKI-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or flakes or pellets
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)62.0-69.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only