Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Fisher BioReagents™

Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™

Catalog NumberQuantityPackaging
A452SK44 LGlass Bottle
Catalog number A452SK4
Price (JPY)
-
Contact Us ›
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Absorbance0.025 max. at 254nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
Assay Percent Range≥99.9 %
Boiling Point64.7°C
CAS67-56-1
ChEBICHEBI:17790
View more
Methanol (Peroxide-Free/Sequencing)
Methanol is used in staining and destaining protein electrophoresis gels, in HPLC, and in other biological applications.