Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Fisher Chemical™

Isopropanol, Optima™LC/MS Grade, Fisher Chemical™

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, isopropanol, 2-hydroxypropane, isopropyl alcohol, alkolave, hartosol, dimethylcarbinol, avantine, sec-propyl alcohol, petrohol, 2-propanol, 2.5L, Report AU at 254nm, Report AU at 230nm, Report AU at 210nm, Report AU at 220nm | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Quantity:
1 L
2.5 L
4 L
Packaging:
View all
Wide Neck Amber Bottle
Catalog number A461-1
Price (JPY)
-
Quantity:
1 L
Packaging:
Wide Neck Amber Bottle
Request bulk or custom format
Specifications
AbsorbanceReport AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
Assay Percent RangeAssay: ≥99.90%
Boiling Point81°C
CAS67-63-0
ChEBICHEBI:17824
View more

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm