Fetal Bovine Serum, Value
Gibco™

Fetal Bovine Serum, Value

Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.
Catalog NumberQuantityFormatHeat InactivatedGamma-irradiated
A520970210 x 50 mLOne ShotYesYes
A520940150 mLOne ShotNoNo
A520940210 x 50 mLOne ShotNoNo
A520950150 mLOne ShotYesNo
A520950210 x 50 mLOne ShotYesNo
A520960150 mLOne ShotNoYes
A520960210 x 50 mLOne ShotNoYes
A520970150 mLOne ShotYesYes
A5256501500 mLBottleNoYes
A5256701500 mLBottleNoNo
A5256801500 mLBottleYesNo
A5256901500 mLBottleYesYes
Catalog number A5209702
Price (JPY)
-
Quantity:
10 x 50 mL
Format:
One Shot
Heat Inactivated:
Yes
Gamma-irradiated:
Yes

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Value

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <20 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
For Use With (Application)*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Gamma-irradiatedYes
Heat InactivatedYes
ProcessingTriple filtered at 0.1 μm
Purity or Quality GradeValue
USDA Safety Testing IncludedUSDA Safety Testing Performed for Australian origins
AgeFetal
Country of OriginRefer to CoA for specific origin*
FormFrozen
FormatOne Shot
Product TypeFetal Bovine Serum
Quantity10 x 50 mL
Serum TreatmentHeat-inact, Gamma-irr
SterilitySterile
Sterilization MethodTriple-filtered, 0.1 μm
Unit SizeEach