Specifications
Absorbance0.005 AU max. at 254nm, 0.02 AU max. at 220nm, 0.04 AU max. at 210nm, 0.05 AU max. at 205nm, 0.07 AU max. at 200nm, 0.15 AU max. at 195nm, 1.00 AU max. at 190nm
Assay Percent RangeAssay: 99.9% min by GC
Boiling Point81°C
CAS75-05-8
ChEBICHEBI:38472
View more