Benzoic Acid, 99.5+%, Extra Pure, SLR, Fisher Chemical™
Benzoic Acid, 99.5+%, Extra Pure, SLR, Fisher Chemical™
Benzoic Acid, 99.5+%, Extra Pure, SLR, Fisher Chemical™
Fisher Chemical™

Benzoic Acid, 99.5+%, Extra Pure, SLR, Fisher Chemical™

Catalog NumberQuantity
B-1900-53
also known as B/1900/53
500 g
Catalog number B-1900-53
also known as B/1900/53
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Specifications
CAS65-85-0
ColorWhite
Formula Weight122.12g/mol
MDL Number2398
Melting Point121°C
View more

Benzoic Acid, 99.5+%, Extra Pure, SLR