Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™
Fisher Chemical™

Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™

Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid
Catalog NumberQuantity
B-4765-77
also known as B/4765/77
50 Tablets
Catalog number B-4765-77
also known as B/4765/77
Price (JPY)
-
Contact Us ›
Quantity:
50 Tablets
Request bulk or custom format
Specifications
ColorWhite
FormulationDisodium Orthophosphate Heptahydrate, Sodium Chloride, Dipotassium Phosphate
MDL Number81857
PackagingGlass Jar
Solubility InformationSoluble
View more
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid