Buffer Tablets, pH 9.2 (Borate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™
Fisher Chemical™

Buffer Tablets, pH 9.2 (Borate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™

Buffer Tablets, pH 9.2 (Borate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid
Catalog NumberQuantity
B-4770-77
also known as B/4770/77
50 Tablets
Catalog number B-4770-77
also known as B/4770/77
Price (JPY)
-
Quantity:
50 Tablets
Request bulk or custom format
Specifications
ColorWhite
FormulationBorates, Sodium Salts, Anhydrous, Sodium Chloride, Tetra
MDL Number81857
PackagingGlass Jar
Solubility InformationSoluble in Water
View more
Buffer Tablets, pH 9.2 (Borate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid