N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™
N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™
N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™
Fisher Chemical™

N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™

Catalog NumberQuantityPackaging
B-4950-08
also known as B/4950/08
500 mLAmber glass bottle
Catalog number B-4950-08
also known as B/4950/08
Price (JPY)
-
Quantity:
500 mL
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS123-86-4
ColorColorless
Formula Weight116.16g/mol
IUPAC Namebutyl acetate
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
View more
N-Butyl Acetate, Extra Pure, SLR