5,6-Dimethyl-2,1,3-benzothiadiazole, 97%
5,6-Dimethyl-2,1,3-benzothiadiazole, 97%
5,6-Dimethyl-2,1,3-benzothiadiazole, 97%
Thermo Scientific Chemicals

5,6-Dimethyl-2,1,3-benzothiadiazole, 97%

CAS: 1887-60-1 | C8H8N2S | 164.226 g/mol
Quantity:
1 g
5 g
Catalog number B20755.03
also known as B20755-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1887-60-1
IUPAC Name5,6-dimethyl-2,1,3-benzothiadiazole
Molecular FormulaC8H8N2S
InChI KeyQQCMLCGOLBUDJF-UHFFFAOYSA-N
SMILESCC1=CC2=NSN=C2C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)78.0-87.0?C
Appearance (Color)Orange to pink
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only