2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid, 97%
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid, 97%
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid, 97%
Thermo Scientific Chemicals

2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid, 97%

CAS: 175201-51-1 | C9H9N3O2 | 191.19 g/mol
Quantity:
1 g
5 g
Catalog number B20771.06
also known as B20771-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS175201-51-1
IUPAC Name(7R)-4,7-dimethyl-4H,7H-pyrazolo[1,5-a]pyrimidine-3-carboxylate
Molecular FormulaC9H10N3O2
InChI KeyNDSVSMFAGULPFP-ZCFIWIBFSA-M
SMILESC[C@@H]1C=CN(C)C2=C(C=NN12)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder or voluminous powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only