3-Chloro-4'-fluoropropiophenone, 97%
3-Chloro-4'-fluoropropiophenone, 97%
3-Chloro-4'-fluoropropiophenone, 97%
3-Chloro-4'-fluoropropiophenone, 97%
Thermo Scientific Chemicals

3-Chloro-4'-fluoropropiophenone, 97%

CAS: 347-93-3 | C9H8ClFO | 186.61 g/mol
Quantity:
5 g
25 g
100 g
Catalog number B21891.14
also known as B21891-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS347-93-3
IUPAC Name3-chloro-1-(4-fluorophenyl)propan-1-one
Molecular FormulaC9H8ClFO
InChI KeyAAHQPLJUSLMHHR-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(=O)CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)43.0-52.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only