Thermo Scientific Chemicals

Diphenyl sulfoxide, 98+%, Thermo Scientific Chemicals

Catalog number: B22135.22
100 g, Each
Thermo Scientific Chemicals

Diphenyl sulfoxide, 98+%, Thermo Scientific Chemicals

Catalog number: B22135.22
100 g, Each
Quantity
Catalog number: B22135.22
also known as B22135-22
Price (JPY)

Chemical Identifiers

CAS
945-51-7
IUPAC Name
(benzenesulfinyl)benzene
Molecular Formula
C12H10OS
InChI Key
JJHHIJFTHRNPIK-UHFFFAOYSA-N
SMILES
O=S(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
67.0-74.0?C

Description

Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.

Solubility
Soluble in water. Soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text