5-Chloro-2-methoxyphenyl isocyanate, 97%
Thermo Scientific Chemicals

5-Chloro-2-methoxyphenyl isocyanate, 97%

Catalog NumberQuantity
B22395.88
also known as B22395-88
2.5 g
Catalog number B22395.88
also known as B22395-88
Price (JPY)
-
Quantity:
2.5 g
Request bulk or custom format
Chemical Identifiers
CAS55440-54-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormFused solid
Assay (GC)≥96.0%
Melting Point (clear melt)53.0-63.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.