Methyl 3-methoxy-4-methylbenzoate, 98%
Methyl 3-methoxy-4-methylbenzoate, 98%
Methyl 3-methoxy-4-methylbenzoate, 98%
Thermo Scientific Chemicals

Methyl 3-methoxy-4-methylbenzoate, 98%

CAS: 3556-83-0 | C10H12O3 | 180.203 g/mol
Quantity:
5 g
25 g
Catalog number B22695.14
also known as B22695-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3556-83-0
IUPAC Namemethyl 3-methoxy-4-methylbenzoate
Molecular FormulaC10H12O3
InChI KeyLLEXCSBUSVRBCA-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentPlaceholder record added to rectify the version number
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only