Di-p-toluoyl-L-tartaric acid, 98%
Di-p-toluoyl-L-tartaric acid, 98%
Di-p-toluoyl-L-tartaric acid, 98%
Thermo Scientific Chemicals

Di-p-toluoyl-L-tartaric acid, 98%

CAS: 32634-66-5 | C20H18O8 | 386.36 g/mol
Quantity:
25 g
100 g
Catalog number B22972.22
also known as B22972-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS32634-66-5
IUPAC Name(2R,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Molecular FormulaC20H18O8
InChI KeyCMIBUZBMZCBCAT-IYBDPMFKSA-N
SMILESCC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Optical Rotation-139.5 ± 2.0? (C=1 in ethanol)
FormPowder
Melting Point (clear melt)164.0-173.0?C
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only