1-Ethynylcyclopentanol, 98+%
1-Ethynylcyclopentanol, 98+%
1-Ethynylcyclopentanol, 98+%
1-Ethynylcyclopentanol, 98+%
Thermo Scientific Chemicals

1-Ethynylcyclopentanol, 98+%

CAS: 17356-19-3 | C7H10O | 110.156 g/mol
Quantity:
5 g
25 g
Catalog number B23109.14
also known as B23109-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS17356-19-3
IUPAC Name1-ethynylcyclopentan-1-ol
Molecular FormulaC7H10O
InChI KeyLQMDOONLLAJAPZ-UHFFFAOYSA-N
SMILESOC1(CCCC1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4720-1.4760 @ 20?C
1-Ethynylcyclopentanol is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylcyclopentanol is used in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Zai-Lai Xie, Qiao-Hua Wei, Li-Yi Zhang, Zhong-Ning Chen. Luminescent Au3Ag6 heterononanuclear complex of 1-ethynylcyclopentanol. Inorganic Chemistry Communications. 2007, 10, (10), 1206-1209.
  2. Vaclav Janout; Steven L. Regen. Phenylmercuric hydroxide. A highly selective reagent for the hydration of nonconjugated terminal alkynes. J. Org. Chem. 1982, 47 (17), 3331-3333.