2-Chloroethyl phenyl sulfide, 98%
2-Chloroethyl phenyl sulfide, 98%
2-Chloroethyl phenyl sulfide, 98%
Thermo Scientific Chemicals

2-Chloroethyl phenyl sulfide, 98%

CAS: 5535-49-9 | C8H9ClS | 172.67 g/mol
Quantity:
5 g
25 g
Catalog number B23265.14
also known as B23265-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5535-49-9
IUPAC Name[(2-chloroethyl)sulfanyl]benzene
Molecular FormulaC8H9ClS
InChI KeyQDXIHHOPZFCEAP-UHFFFAOYSA-N
SMILESClCCSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5815-1.5855 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only