L-Leucinol, 97%
L-Leucinol, 97%
L-Leucinol, 97%
Thermo Scientific Chemicals

L-Leucinol, 97%

CAS: 7533-40-6 | C6H15NO | 117.19 g/mol
Quantity:
5 g
25 g
Catalog number B23745.14
also known as B23745-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7533-40-6
IUPAC Name(2S)-2-amino-4-methylpentan-1-ol
Molecular FormulaC6H15NO
InChI KeyVPSSPAXIFBTOHY-LURJTMIESA-N
SMILESCC(C)C[C@H](N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Optical Rotation1.4 ± 0.2? (neat)
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid or viscous liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only