4-Benzyloxy-2-hydroxypyridine, 97%
4-Benzyloxy-2-hydroxypyridine, 97%
4-Benzyloxy-2-hydroxypyridine, 97%
4-Benzyloxy-2-hydroxypyridine, 97%
Thermo Scientific Chemicals

4-Benzyloxy-2-hydroxypyridine, 97%

CAS: 53937-02-3 | C12H11NO2 | 201.23 g/mol
Quantity:
5 g
25 g
Catalog number B24774.06
also known as B24774-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS53937-02-3
IUPAC Name4-(benzyloxy)-1,2-dihydropyridin-2-one
Molecular FormulaC12H11NO2
InChI KeyDOVNUEPFPBWTSV-UHFFFAOYSA-N
SMILESO=C1NC=CC(OCC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only