Thermo Scientific Chemicals

4,4'-Dimethylbiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: B25089.03
1 g, Each
Thermo Scientific Chemicals

4,4'-Dimethylbiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: B25089.03
1 g, Each
Quantity
Catalog number: B25089.03
also known as B25089-03
Price (JPY)

Chemical Identifiers

CAS
613-33-2
IUPAC Name
4,4'-dimethyl-1,1'-biphenyl
Molecular Formula
C14H14
InChI Key
RZTDESRVPFKCBH-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C1=CC=C(C)C=C1
Appearance (Color)
White to cream or pale yellow
Form
Crystals, crystalline powder or flakes
Assay (GC)
≥98.5%
Melting Point
119-125?C

Description

4,4'-Dimethylbiphenyl is used in the preparation of biphenyl-4,4’-dicarboxylic acid, which acts as a monomer utilized in the synthesis of linear long chain polymer through polycondensation reaction. It is used to prepare 4-(4'-methylphenyl)benzaldehyde by selective photoxygenation reaction using 9-phenyl-10-methylacridinium perchlorate as a catalyst under visible light irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbiphenyl is used in the preparation of biphenyl-4,4’-dicarboxylic acid, which acts as a monomer utilized in the synthesis of linear long chain polymer through polycondensation reaction. It is used to prepare 4-(4′-methylphenyl)benzaldehyde by selective photoxygenation reaction using 9-phenyl-10-methylacridinium perchlorate as a catalyst under visible light irradiation.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text