Specifications
CAS1239-45-8
CAS7732-18-5
ChEBICHEBI:4883
ChemAlert Storage SymbolBlue
Chemical Name or MaterialEthidium Bromide
ColorRed
Formula Weight394.33
IUPAC Name5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
InChI KeyZMMJGEGLRURXTF-UHFFFAOYSA-N
MDL NumberMFCD00011724
Merck Index15, 4769
Molecular FormulaC21H20BrN3
Name Note1% Solution
PackagingClear Bottle
Physical FormLiquid
PubChem CID14710
Purity Grade NotesDNase-, RNase- and Protease-Free
Recommended StorageRT
SMILES[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Synonymethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
GradeMolecular Biology
Molecular Weight (g/mol)394.32
Quantity10 mL
Unit SizeEach