Agar, Powder / Flakes, Fisher BioReagents™
Actual product may vary
Fisher BioReagents™

Agar, Powder / Flakes, Fisher BioReagents™

Used as a solidifying agent for culture media.
Catalog NumberQuantityPackaging Type
BP1423-22000 gPoly Pail
Catalog number BP1423-2
Price (JPY)
-
Quantity:
2000 g
Packaging Type:
Poly Pail
Used as a solidifying agent for culture media
  • Used as a solidfying agent to provide the solid medium to support the growth of microbes for cell culture applications
  • Functionally tested for promotion of growth of E. coli and S. aureus
  • Typical concentration is 1.5% agar (15g/L) in water
  • CAS: 9002-18-0
  • Molecular Weight: 336.337 g/mol
  • Chemical Formula: C14H24O9
Specifications
For Use With (Application)Used for a solidifying agent for culture media
GradeMolecular genetics
Packaging TypePoly Pail
Quantity2000 g
FormPowder
Product TypeAgar
Unit SizeEach